Assay Details
| Gene Symbol: GluProRS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079747.3 | NP_524471.2 | 2 - 3 | 1070 | 63 | |
| GenBank mRNA | AY058703.1 | - | 2 - 3 | 1046 | 63 | |
More Information
Set Membership: |
|

GluProRS
Genomic Map