Assay Details
| Gene Symbol: Rab7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260347.1 | NP_001247276.1 | 2 - 3 | 324 | 103 | |
| NM_001260348.2 | NP_001247277.1 | 2 - 3 | 335 | 103 | ||
| NM_079748.4 | NP_524472.1 | 2 - 3 | 370 | 103 | ||
| GenBank mRNA | AB035672.1 | - | 2 - 3 | 356 | 103 | |
| AF263363.1 | - | 2 - 3 | 350 | 103 | ||
| AY060236.1 | - | 2 - 3 | 350 | 103 | ||
| BT124755.1 | - | 2 - 3 | 324 | 103 | ||
More Information
Set Membership: |
|

Rab7
Genomic Map