Assay Details
| Gene Symbol: Syx1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316616.1 | NP_001303545.1 | 1 | 2156 | 111 | |
| NM_079751.5 | NP_524475.1 | 1 | 2156 | 111 | ||
| GenBank mRNA | AY061472.1 | - | 1 | 2136 | 111 | |
| L37732.1 | - | 1 | 1129 | 111 | ||
More Information
Set Membership: |
|

Syx1A
Genomic Map