Assay Details
| Gene Symbol: Kaz-m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276051.2 | NP_001262980.1 | 1 - 2 | 304 | 85 | |
| NM_079783.4 | NP_524507.1 | 1 - 2 | 304 | 85 | ||
| GenBank mRNA | AY113480.1 | - | 1 - 2 | 301 | 85 | |
| KX531894.1 | - | 1 - 2 | 209 | 85 | ||
More Information
Set Membership: |
|

Kaz-m1
Genomic Map