Assay Details
| Gene Symbol: Rb97D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014671.2 | NP_001014671.1 | 4 - 5 | 1398 | 75 | |
| NM_001014672.2 | NP_001014672.1 | 4 - 5 | 1398 | 75 | ||
| NM_001014674.2 | NP_001014674.1 | 4 - 5 | 1398 | 75 | ||
| NM_079796.5 | NP_524520.1 | 4 - 5 | 1398 | 75 | ||
More Information
Set Membership: |
|

Rb97D
Genomic Map