Assay Details
| Gene Symbol: l(3)mbt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170330.2 | NP_733209.1 | 4 - 5 | 3024 | 60 | |
| GenBank mRNA | AY051653.1 | - | 4 - 5 | 3024 | 60 | |
More Information
Set Membership: |
|

l(3)mbt
Genomic Map