Assay Details
| Gene Symbol: Mtl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079809.3 | NP_524533.1 | 2 - 3 | 456 | 73 | |
| NM_170343.2 | NP_733222.1 | 2 - 3 | 361 | 73 | ||
| NM_170344.2 | NP_733223.1 | 2 - 3 | 332 | 73 | ||
| GenBank mRNA | AF238044.1 | - | 2 - 3 | 403 | 73 | |
| AJ413188.1 | - | 2 - 3 | 403 | 73 | ||
| AY113279.1 | - | 2 - 3 | 332 | 73 | ||
More Information
Set Membership: |
|

Mtl
Genomic Map