Assay Details
| Gene Symbol: Mst98Cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079815.3 | NP_524539.1 | 1 | 1001 | 67 | |
| GenBank mRNA | AY118617.1 | - | 1 | 983 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mst98Cb| Gene Symbol: Mst98Cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079815.3 | NP_524539.1 | 1 | 1001 | 67 | |
| GenBank mRNA | AY118617.1 | - | 1 | 983 | 67 | |
Set Membership: |
|