Assay Details
| Gene Symbol: Hrb98DE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079819.3 | NP_524543.1 | 3 - 4 | 831 | 66 | |
| NM_170370.2 | NP_733249.1 | 3 - 4 | 835 | 66 | ||
| NM_170371.2 | NP_733250.1 | 3 - 4 | 823 | 66 | ||
| NM_170372.2 | NP_733251.1 | 3 - 4 | 831 | 66 | ||
| NM_170373.2 | NP_733252.1 | 3 - 4 | 819 | 66 | ||
| NM_170374.2 | NP_733253.1 | 3 - 4 | 819 | 66 | ||
| GenBank mRNA | AY061448.1 | - | 3 - 4 | 822 | 66 | |
| BT099813.1 | - | 3 - 4 | 821 | 66 | ||
| M15766.1 | - | 3 - 4 | 880 | 66 | ||
More Information
Set Membership: |
|

Hrb98DE
Genomic Map