Assay Details
| Gene Symbol: Pkc98E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300647.1 | NP_001287576.1 | 6 - 7 | 976 | 71 | |
| NM_001300648.1 | NP_001287577.1 | 6 - 7 | 1010 | 71 | ||
| NM_079821.4 | NP_524545.2 | 6 - 7 | 976 | 71 | ||
More Information
Set Membership: |
|

Pkc98E
Genomic Map