Assay Details
| Gene Symbol: Dop1R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276143.1 | NP_001263072.1 | 3 - 4 | 2357 | 94 | |
| NM_079824.4 | NP_524548.1 | 3 - 4 | 2357 | 94 | ||
| NM_170420.2 | NP_733299.1 | 3 - 4 | 2357 | 94 | ||
| GenBank mRNA | U34383.1 | - | 1 - 2 | 1561 | 94 | |
| U61264.1 | - | 2 - 3 | 2048 | 94 | ||
More Information
Set Membership: |
|

Dop1R2
Genomic Map