Assay Details
| Gene Symbol: Dop1R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079824.4 | NP_524548.1 | 5 - 6 | 2607 | 59 | |
| GenBank mRNA | U34383.1 | - | 3 - 4 | 1811 | 59 | |
More Information
Set Membership: |
|

Dop1R2
Genomic Map