Assay Details
Gene Symbol: TkR99D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170301.1 | NP_001163772.1 | 6 - 7 | 1576 | 64 | |
NM_001276163.1 | NP_001263092.1 | 6 - 7 | 1576 | 64 | ||
NM_079832.3 | NP_524556.2 | 6 - 7 | 1576 | 64 | ||
GenBank mRNA | BT150106.1 | - | 6 - 7 | 1576 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
