Assay Details
Gene Symbol: TkR99D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276163.1 | NP_001263092.1 | 9 - 10 | 1981 | 71 | |
NM_079832.3 | NP_524556.2 | 9 - 10 | 1981 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
