Assay Details
| Gene Symbol: PH4alphaMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079855.3 | NP_524594.2 | 4 - 5 | 1401 | 66 | |
| GenBank mRNA | BT132909.1 | - | 4 - 5 | 1400 | 66 | |
More Information
Set Membership: |
|

PH4alphaMP
Genomic Map