Assay Details
| Gene Symbol: Gycbeta100B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079864.4 | NP_524603.2 | 3 - 4 | 448 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gycbeta100B| Gene Symbol: Gycbeta100B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079864.4 | NP_524603.2 | 3 - 4 | 448 | 74 | |
Set Membership: |
|