Assay Details
| Gene Symbol: gwl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080121.4 | NP_524860.2 | 3 - 4 | 1344 | 72 | |
| GenBank mRNA | AY058648.1 | - | 3 - 4 | 1316 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gwl| Gene Symbol: gwl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080121.4 | NP_524860.2 | 3 - 4 | 1344 | 72 | |
| GenBank mRNA | AY058648.1 | - | 3 - 4 | 1316 | 72 | |
Set Membership: |
|