Assay Details
| Gene Symbol: gwl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275784.1 | NP_001262713.1 | 4 - 5 | 2353 | 82 | |
| NM_080121.4 | NP_524860.2 | 5 - 6 | 2482 | 82 | ||
| GenBank mRNA | AY058648.1 | - | 5 - 6 | 2454 | 82 | |
More Information
Set Membership: |
|

gwl
Genomic Map