Assay Details
| Gene Symbol: Mst57Da | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080167.4 | NP_524906.1 | 1 | 84 | 67 | |
| GenBank mRNA | KX531385.1 | - | 1 | 61 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mst57Da| Gene Symbol: Mst57Da | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080167.4 | NP_524906.1 | 1 | 84 | 67 | |
| GenBank mRNA | KX531385.1 | - | 1 | 61 | 67 | |
Set Membership: |
|