Assay Details
| Gene Symbol: krz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260471.2 | NP_001247400.1 | 1 - 2 | 720 | 94 | |
| NM_080249.5 | NP_524988.1 | 1 - 2 | 720 | 94 | ||
| GenBank mRNA | AY118946.1 | - | 1 - 2 | 655 | 94 | |
More Information
Set Membership: |
|

krz
Genomic Map