Assay Details
| Gene Symbol: bor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260206.1 | NP_001247135.1 | 5 - 6 | 1832 | 74 | |
| NM_080257.4 | NP_524996.1 | 4 - 5 | 1865 | 74 | ||
| GenBank mRNA | AF227209.1 | - | 4 - 5 | 1849 | 74 | |
More Information
Set Membership: |
|

bor
Genomic Map