Assay Details
| Gene Symbol: larp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170286.3 | NP_001163757.1 | 4 - 5 | 1582 | 72 | |
| NM_001260417.2 | NP_001247346.1 | 4 - 5 | 1832 | 72 | ||
| NM_001260418.2 | NP_001247347.1 | 5 - 6 | 2677 | 72 | ||
| NM_001260419.2 | NP_001247348.1 | 5 - 6 | 2681 | 72 | ||
| NM_080259.3 | NP_524998.1 | 4 - 5 | 1507 | 72 | ||
| NM_170365.4 | NP_733244.5 | 5 - 6 | 2826 | 72 | ||
| NM_170366.4 | NP_733245.1 | 4 - 5 | 1519 | 72 | ||
| GenBank mRNA | BT128788.1 | - | 1 - 2 | 80 | 72 | |
More Information
Set Membership: |
|

larp
Genomic Map