Assay Details
| Gene Symbol: larp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170286.3 | NP_001163757.1 | 5 - 6 | 3697 | 60 | |
| NM_001260417.2 | NP_001247346.1 | 5 - 6 | 3947 | 60 | ||
| NM_001260418.2 | NP_001247347.1 | 6 - 7 | 4792 | 60 | ||
| NM_001260419.2 | NP_001247348.1 | 6 - 7 | 4796 | 60 | ||
| NM_080259.3 | NP_524998.1 | 5 - 6 | 3622 | 60 | ||
| NM_170365.4 | NP_733244.5 | 6 - 7 | 4941 | 60 | ||
| NM_170366.4 | NP_733245.1 | 5 - 6 | 3634 | 60 | ||
| GenBank mRNA | BT128788.1 | - | 2 - 3 | 2195 | 60 | |
More Information
Set Membership: |
|

larp
Genomic Map