Assay Details
| Gene Symbol: DNApol-alpha180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080488.3 | NP_536736.2 | 3 - 4 | 1340 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DNApol-alpha180| Gene Symbol: DNApol-alpha180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080488.3 | NP_536736.2 | 3 - 4 | 1340 | 71 | |
Set Membership: |
|