Assay Details
| Gene Symbol: NPF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275713.1 | NP_001262642.1 | 2 - 3 | 307 | 120 | |
| NM_001275714.1 | NP_001262643.1 | 2 - 3 | 368 | 120 | ||
| NM_080493.3 | NP_536741.1 | 2 - 3 | 358 | 120 | ||
| GenBank mRNA | BT023797.1 | - | 2 - 3 | 358 | 120 | |
More Information
Set Membership: |
|

NPF
Genomic Map