Assay Details
| Gene Symbol: Zip89B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080499.3 | NP_536747.2 | 1 - 2 | 1643 | 68 | |
| GenBank mRNA | BT133201.1 | - | 1 - 2 | 1638 | 68 | |
More Information
Set Membership: |
|

Zip89B
Genomic Map