Assay Details
| Gene Symbol: modSP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080515.3 | NP_536776.2 | 4 - 5 | 1319 | 72 | |
| GenBank mRNA | AY118964.1 | - | 4 - 5 | 1424 | 72 | |
More Information
Set Membership: |
|

modSP
Genomic Map