Assay Details
| Gene Symbol: spn-D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170321.3 | NP_733200.1 | 5 - 6 | 722 | 55 | |
| GenBank mRNA | AY071162.1 | - | 5 - 6 | 720 | 55 | |
| AY257540.1 | - | 5 - 6 | 718 | 55 | ||
More Information
Set Membership: |
|

spn-D
Genomic Map