Assay Details
| Gene Symbol: BOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170280.2 | NP_001163751.1 | 3 - 4 | 3453 | 69 | |
| NM_170361.3 | NP_733240.1 | 3 - 4 | 3453 | 69 | ||
| GenBank mRNA | BT032836.1 | - | 3 - 4 | 3384 | 69 | |
More Information
Set Membership: |
|

BOD1
Genomic Map