Assay Details
| Gene Symbol: Hrb98DE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170370.2 | NP_733249.1 | 5 - 6 | 1229 | 134 | |
| NM_170371.2 | NP_733250.1 | 5 - 6 | 1217 | 134 | ||
| NM_170372.2 | NP_733251.1 | 5 - 6 | 1225 | 134 | ||
| NM_170373.2 | NP_733252.1 | 5 - 6 | 1213 | 134 | ||
| GenBank mRNA | AY061448.1 | - | 5 - 6 | 1216 | 134 | |
| BT099813.1 | - | 5 - 6 | 1215 | 134 | ||
More Information
Set Membership: |
|

Hrb98DE
Genomic Map