Assay Details
| Gene Symbol: spg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144646.2 | NP_001138118.2 | 6 - 7 | 949 | 61 | |
| NM_001276118.1 | NP_001263047.1 | 6 - 7 | 919 | 61 | ||
| NM_001276119.1 | NP_001263048.1 | 5 - 6 | 1458 | 61 | ||
| NM_001276120.1 | NP_001263049.1 | 5 - 6 | 1458 | 61 | ||
| NM_170379.5 | NP_733258.4 | 6 - 7 | 919 | 61 | ||
More Information
Set Membership: |
|

spg
Genomic Map