Assay Details
| Gene Symbol: alph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260427.2 | NP_001247356.1 | 7 - 8 | 1729 | 77 | |
| NM_170419.3 | NP_733298.1 | 6 - 7 | 1645 | 77 | ||
More Information
Set Membership: |
|

alph
Genomic Map