Assay Details
| Gene Symbol: kay | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032406.2 | NP_001027578.1 | 1 - 2 | 540 | 94 | |
| GenBank mRNA | AY069805.1 | - | 1 - 2 | 517 | 94 | |
| DQ858476.1 | - | 1 - 2 | 538 | 94 | ||
More Information
Set Membership: |
|

kay
Genomic Map