Assay Details
| Gene Symbol: ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300664.1 | NP_001287593.1 | 4 - 5 | 2036 | 61 | |
| NM_170430.3 | NP_733309.3 | 4 - 5 | 2463 | 61 | ||
| GenBank mRNA | BT057978.1 | - | 4 - 5 | 2026 | 61 | |
More Information
Set Membership: |
|

ca
Genomic Map