Assay Details
| Gene Symbol: ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300664.1 | NP_001287593.1 | 7 - 8 | 3198 | 118 | |
| NM_170430.3 | NP_733309.3 | 7 - 8 | 3625 | 118 | ||
| GenBank mRNA | BT057978.1 | - | 7 - 8 | 3188 | 118 | |
More Information
Set Membership: |
|

ca
Genomic Map