Assay Details
| Gene Symbol: Atg16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144652.2 | NP_001138124.2 | 7 - 8 | 1769 | 150 | |
| NM_170432.3 | NP_733311.2 | 5 - 6 | 1054 | 150 | ||
| NM_170433.3 | NP_733312.2 | 4 - 5 | 1063 | 150 | ||
| NM_170434.4 | NP_733313.2 | 7 - 8 | 1745 | 150 | ||
| GenBank mRNA | AY058742.1 | - | 5 - 6 | 1054 | 150 | |
| BT024267.1 | - | 7 - 8 | 1695 | 150 | ||
More Information
Set Membership: |
|

Atg16
Genomic Map