Assay Details
| Gene Symbol: Cad99C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202382.1 | NP_001189311.1 | 4 - 5 | 598 | 83 | |
| NM_170435.2 | NP_733314.1 | 5 - 6 | 747 | 83 | ||
More Information
Set Membership: |
|

Cad99C
Genomic Map