Assay Details
| Gene Symbol: Cad99C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202382.1 | NP_001189311.1 | 9 - 10 | 4398 | 68 | |
| NM_170435.2 | NP_733314.1 | 10 - 11 | 4547 | 68 | ||
More Information
Set Membership: |
|

Cad99C
Genomic Map