Assay Details
| Gene Symbol: Axn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144654.3 | NP_001138126.1 | 4 - 5 | 1554 | 74 | |
| NM_001170302.1 | NP_001163773.1 | 2 - 3 | 501 | 74 | ||
| NM_001276168.1 | NP_001263097.1 | 4 - 5 | 1554 | 74 | ||
| NM_001276169.1 | NP_001263098.1 | 2 - 3 | 501 | 74 | ||
| NM_170457.3 | NP_733336.1 | 4 - 5 | 1554 | 74 | ||
| NM_170458.2 | NP_733337.1 | 2 - 3 | 501 | 74 | ||
| NM_170459.3 | NP_733338.1 | 4 - 5 | 1554 | 74 | ||
| GenBank mRNA | AF091813.1 | - | 4 - 5 | 824 | 74 | |
| BT133166.1 | - | 1 - 2 | 197 | 74 | ||
More Information
Set Membership: |
|

Axn
Genomic Map