Assay Details
| Gene Symbol: Axn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144654.3 | NP_001138126.1 | 7 - 8 | 2570 | 64 | |
| NM_001170302.1 | NP_001163773.1 | 4 - 5 | 1595 | 64 | ||
| NM_001276168.1 | NP_001263097.1 | 7 - 8 | 2570 | 64 | ||
| NM_001276169.1 | NP_001263098.1 | 6 - 7 | 1655 | 64 | ||
| NM_170457.3 | NP_733336.1 | 8 - 9 | 2708 | 64 | ||
| NM_170458.2 | NP_733337.1 | 6 - 7 | 1655 | 64 | ||
| NM_170459.3 | NP_733338.1 | 8 - 9 | 2708 | 64 | ||
| GenBank mRNA | AF091813.1 | - | 8 - 9 | 1978 | 64 | |
| BT133166.1 | - | 4 - 5 | 1213 | 64 | ||
More Information
Set Membership: |
|

Axn
Genomic Map