Assay Details
| Gene Symbol: Axn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144654.3 | NP_001138126.1 | 8 - 9 | 2704 | 70 | |
| NM_001170302.1 | NP_001163773.1 | 5 - 6 | 1729 | 70 | ||
| NM_001276168.1 | NP_001263097.1 | 8 - 9 | 2704 | 70 | ||
| NM_001276169.1 | NP_001263098.1 | 7 - 8 | 1789 | 70 | ||
| NM_170457.3 | NP_733336.1 | 9 - 10 | 2842 | 70 | ||
| NM_170458.2 | NP_733337.1 | 7 - 8 | 1789 | 70 | ||
| NM_170459.3 | NP_733338.1 | 9 - 10 | 2842 | 70 | ||
| GenBank mRNA | AF091813.1 | - | 9 - 10 | 2112 | 70 | |
| BT133166.1 | - | 5 - 6 | 1347 | 70 | ||
More Information
Set Membership: |
|

Axn
Genomic Map