Assay Details
| Gene Symbol: CG31028 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170473.3 | NP_733352.3 | 7 - 8 | 3017 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CG31028| Gene Symbol: CG31028 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170473.3 | NP_733352.3 | 7 - 8 | 3017 | 68 | |
Set Membership: |
|