Assay Details
| Gene Symbol: PH4alphaEFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170492.2 | NP_733371.1 | 5 - 6 | 1189 | 54 | |
| GenBank mRNA | AF495536.1 | - | 5 - 6 | 1174 | 54 | |
| AY052109.1 | - | 5 - 6 | 1174 | 54 | ||
More Information
Set Membership: |
|

PH4alphaEFB
Genomic Map