Assay Details
| Gene Symbol: PH4alphaNE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170495.2 | NP_733374.1 | 5 - 6 | 948 | 92 | |
| GenBank mRNA | BT132956.1 | - | 5 - 6 | 948 | 92 | |
More Information
Set Membership: |
|

PH4alphaNE1
Genomic Map