Assay Details
| Gene Symbol: PH4alphaPV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170516.1 | NP_733395.1 | 4 - 5 | 889 | 71 | |
| GenBank mRNA | AF495539.1 | - | 4 - 5 | 889 | 71 | |
| BT150220.1 | - | 4 - 5 | 889 | 71 | ||
More Information
Set Membership: |
|

PH4alphaPV
Genomic Map