Assay Details
| Gene Symbol: PH4alphaPV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170516.1 | NP_733395.1 | 6 - 7 | 1395 | 63 | |
| GenBank mRNA | AF495539.1 | - | 6 - 7 | 1395 | 63 | |
| BT150220.1 | - | 6 - 7 | 1395 | 63 | ||
More Information
Set Membership: |
|

PH4alphaPV
Genomic Map