Assay Details
| Gene Symbol: cindr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276195.1 | NP_001263124.1 | 3 - 4 | 1112 | 83 | |
| NM_001276200.1 | NP_001263129.1 | 3 - 4 | 1032 | 83 | ||
| NM_170526.3 | NP_733405.2 | 3 - 4 | 1032 | 83 | ||
More Information
Set Membership: |
|

cindr
Genomic Map