Assay Details
| Gene Symbol: cindr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170527.3 | NP_733406.1 | 3 - 4 | 1788 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cindr| Gene Symbol: cindr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170527.3 | NP_733406.1 | 3 - 4 | 1788 | 95 | |
Set Membership: |
|