Assay Details
| Gene Symbol: pyd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104242.3 | NP_001097712.1 | 3 - 4 | 1296 | 63 | |
| NM_001260071.2 | NP_001247000.1 | 2 - 3 | 675 | 63 | ||
| NM_170646.6 | NP_731293.3 | 3 - 4 | 1768 | 63 | ||
More Information
Set Membership: |
|

pyd
Genomic Map