Assay Details
| Gene Symbol: CG42724 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202255.1 | NP_001189184.1 | 3 - 4 | 2205 | 65 | |
| NM_001202257.3 | NP_001189186.1 | 3 - 4 | 2106 | 65 | ||
| NM_001275385.1 | NP_001262314.1 | 3 - 4 | 2106 | 65 | ||
| NM_001300264.1 | NP_001287193.1 | 3 - 4 | 2106 | 65 | ||
| NM_001300265.1 | NP_001287194.1 | 3 - 4 | 2106 | 65 | ||
| NM_176408.2 | NP_788585.1 | 3 - 4 | 2106 | 65 | ||
| GenBank mRNA | AY069106.1 | - | 3 - 4 | 2077 | 65 | |
More Information
Set Membership: |
|

CG42724
Genomic Map